CAS RN 263563-09-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MilliporeSigma™ Calbiochem™ β-Secretase Inhibitor II
The β-Secretase Inhibitor II, also referenced under CAS 263563-09-3, controls the biological activity of β-Secretase. This small molecule/inhibitor is primarily used for Neuroscience applications.