accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 263874-05-1

IUPAC Name: 3-methyl-1-[6-(3-methyl-1H-imidazol-3-ium-1-yl)pyridin-2-yl]-1H-imidazol-3-ium dibromide
Synonyms: 1-methyl-3-[6-(3-methylimidazol-3-ium-1-yl)pyridin-2-yl]imidazol-1-ium dibromide
Molecular Weight (g/mol): 401.11
Molecular Formula: C13H15Br2N5
InChi Key: VAESRVWVZJLVIA-UHFFFAOYSA-L
SMILES: [Br-].[Br-].C[N+]1=CN(C=C1)C1=CC=CC(=N1)N1C=C[N+](C)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)