accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNNOOFFFNOO

CAS RN 26399-36-0

IUPAC Name: N-(cyclopropylmethyl)-2,6-dinitro-N-propyl-4-(trifluoromethyl)aniline
Synonyms: profluralin
Molecular Weight (g/mol): 347.29
Molecular Formula: C14H16F3N3O4
InChi Key: ITVQAKZNYJEWKS-UHFFFAOYSA-N
SMILES: CCCN(CC1CC1)C1=C(C=C(C=C1[N+]([O-])=O)C(F)(F)F)[N+]([O-])=O
Grade 
  • (1)
Quantity 
  • (1)
Solvent or Matrix 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results