accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2641-89-6

IUPAC Name: tetrabromobenzene-1,4-diol
Synonyms: 1,4-dihydroxytetrabromobenzene
Molecular Weight (g/mol): 425.7
Molecular Formula: C6H2Br4O2
InChi Key: DTFQULSULHRJOA-UHFFFAOYSA-N
SMILES: OC1=C(Br)C(Br)=C(O)C(Br)=C1Br
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results