accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2644-70-4

IUPAC Name: dihydrogen hydrazine dichloride
Synonyms: dihydrogen hydrazine dichloride
Molecular Weight (g/mol): 104.96
Molecular Formula: Cl2H6N2
InChi Key: LIAWOTKNAVAKCX-UHFFFAOYSA-N
SMILES: [H+].[H+].[Cl-].[Cl-].NN
Molecular Weight (g/mol) 
  • (2)
  • (5)
Percent Purity 
  • (3)
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (2)
  • (3)
  • (2)
  • (5)
  • (3)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)