accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 26446-38-8

IUPAC Name: 2-{[3,4-dihydroxy-2,5-bis(hydroxymethyl)oxolan-2-yl]oxy}-6-(hydroxymethyl)oxane-3,4,5-triol; hexadecanoic acid
Synonyms: granulated sugar; palmitic acid
Molecular Weight (g/mol): 598.73
Molecular Formula: C28H54O13
InChi Key: XZAGBDSOKNXTDT-UHFFFAOYNA-N
SMILES: CCCCCCCCCCCCCCCC(O)=O.OCC1OC(CO)(OC2OC(CO)C(O)C(O)C2O)C(O)C1O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)