accessibility menu, dialog, popup

UserName

Viewing profile as user:
HClH3COONNH2H2NNHO

CAS RN 2645-08-1

IUPAC Name: hydrogen ethyl 5-[(diaminomethylidene)amino]-2-(phenylformamido)pentanoate chloride
Synonyms: hydrogen ethyl 5-[(diaminomethylidene)amino]-2-(phenylformamido)pentanoate chloride
Molecular Weight (g/mol): 342.82
Molecular Formula: C15H23ClN4O3
InChi Key: HIXDELXKSSLIKB-UHFFFAOYNA-N
SMILES: [H+].[Cl-].CCOC(=O)C(CCCN=C(N)N)NC(=O)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (3)
  • (1)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)