accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 26452-98-2

IUPAC Name: 2,3-dihydro-1H-inden-1-yl acetate
Synonyms: 2,3-dihydro-1H-inden-1-yl acetate
Molecular Weight (g/mol): 176.22
Molecular Formula: C11H12O2
InChi Key: BZZWYQKRGRYJAX-UHFFFAOYNA-N
SMILES: CC(=O)OC1CCC2=CC=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results