accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 26456-05-3

IUPAC Name: 10-methylacridin-10-ium perchlorate
Synonyms: 10-methylacridin-10-ium perchlorate
Molecular Weight (g/mol): 293.7
Molecular Formula: C14H12ClNO4
InChi Key: UTTIEKPPJSQKJC-UHFFFAOYSA-M
SMILES: [O-][Cl](=O)(=O)=O.C[N+]1=C2C=CC=CC2=CC2=CC=CC=C12
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results