accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2646-30-2

NH2SNHBr

CAS RN 2646-30-2

IUPAC Name: (4-bromophenyl)thiourea
Synonyms: 4-bromophenyl thiourea C7H7BrN2S
Molecular Weight (g/mol): 231.11
Molecular Formula: C7H7BrN2S
InChi Key: MRVQULNOKCOGHC-UHFFFAOYSA-N
SMILES: NC(=S)NC1=CC=C(Br)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results