accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2648-71-7

H3COCH3CH3Br

CAS RN 2648-71-7

IUPAC Name: 3-bromo-3-methylbutan-2-one
Synonyms: 3-bromo-3-methylbutan-2-one
Molecular Weight (g/mol): 165.03
Molecular Formula: C5H9BrO
InChi Key: XHCWDDGZFBAXRT-UHFFFAOYSA-N
SMILES: CC(=O)C(C)(C)Br
Quantity 
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results