accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 26487-67-2

IUPAC Name: 1-(2-chloroethyl)azepan-1-ium chloride
Synonyms: 1-(2-chloroethyl)azepan-1-ium chloride
Molecular Weight (g/mol): 198.13
Molecular Formula: C8H17Cl2N
InChi Key: ZQDSOUPBYJIPNM-UHFFFAOYSA-N
SMILES: [Cl-].ClCC[NH+]1CCCCCC1
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)