CAS RN 26494-76-8
CAS RN 26494-76-8
IUPAC Name:
4-amino-1λ⁶-thiomorpholine-1,1-dione
Synonyms:
4-amino-1λ⁶-thiomorpholine-1,1-dione
Molecular Weight (g/mol):
150.2
Molecular Formula:
C4H10N2O2S
InChi Key:
YDKNASMIEZGJEJ-UHFFFAOYSA-N
SMILES:
NN1CCS(=O)(=O)CC1
1
–
1
of
1
results
1
4-Aminothiomorpholine 1,1-Dioxide 95.0+%, TCI America™
CAS: 26494-76-8 Molecular Formula: C4H10N2O2S Molecular Weight (g/mol): 150.196 InChI Key: YDKNASMIEZGJEJ-UHFFFAOYSA-N Synonym: 1,1-Dioxo-4-thiomorpholinamine, 4-Thiomorpholinamine 1,1-Dioxide PubChem CID: 12708263 IUPAC Name: 1,1-dioxo-1,4-thiazinan-4-amine SMILES: C1CS(=O)(=O)CCN1N