accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2650-50-2

IUPAC Name: trimethyl(propyl)azanium bromide
Synonyms: nervonyl carnitine bromide
Molecular Weight (g/mol): 182.11
Molecular Formula: C6H16BrN
InChi Key: FCGQIZKUTMUWDC-UHFFFAOYSA-M
SMILES: [Br-].CCC[N+](C)(C)C
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results