accessibility menu, dialog, popup

UserName

Viewing profile as user:
BrH3CNCH3CH3H3C

CAS RN 2650-53-5

IUPAC Name: hexyltrimethylazanium bromide
Synonyms: hexyltrimethylazanium bromide
Molecular Weight (g/mol): 224.19
Molecular Formula: C9H22BrN
InChi Key: JYVPKRHOTGQJSE-UHFFFAOYSA-M
SMILES: [Br-].CCCCCC[N+](C)(C)C
Molecular Weight (g/mol) 
  • (1)
  • (4)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (4)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)