accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 26549-65-5

IUPAC Name: (2R,3S)-2,3-dihydroxy-N,N,N',N'-tetramethylbutanediamide
Synonyms: (2R,3S)-2,3-dihydroxy-N,N,N',N'-tetramethylsuccinamide
Molecular Weight (g/mol): 204.23
Molecular Formula: C8H16N2O4
InChi Key: PCYDYHRBODKVEL-OLQVQODUSA-N
SMILES: CN(C)C(=O)[C@@H](O)[C@@H](O)C(=O)N(C)C
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results