CAS RN 26557-78-8
IUPAC Name:
4-(prop-2-yn-1-yloxy)aniline
Synonyms:
4-(prop-2-yn-1-yloxy)aniline
Molecular Weight (g/mol):
147.18
Molecular Formula:
C9H9NO
InChi Key:
FPLSLCJCSKFAMA-UHFFFAOYSA-N
SMILES:
NC1=CC=C(OCC#C)C=C1
1
–
1
of
1
results
1
4-(2-Propynyloxy)aniline 98.0+%, TCI America™
CAS: 26557-78-8 Molecular Formula: C9H9NO Molecular Weight (g/mol): 147.18 MDL Number: MFCD12823190 InChI Key: FPLSLCJCSKFAMA-UHFFFAOYSA-N Synonym: 4-(Propargyloxy)aniline, 4-Aminophenyl Propargyl Ether PubChem CID: 12668862 IUPAC Name: 4-(prop-2-yn-1-yloxy)aniline SMILES: NC1=CC=C(OCC#C)C=C1