accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 265664-52-6

IUPAC Name: [4-(2-methoxyethoxy)phenyl]boronic acid
Synonyms: 4-(2-methoxyethoxy)phenylboronic acid
Molecular Weight (g/mol): 196.01
Molecular Formula: C9H13BO4
InChi Key: ODKMFCPXFMQWHA-UHFFFAOYSA-N
SMILES: COCCOC1=CC=C(C=C1)B(O)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results