accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2657-87-6

IUPAC Name: 4-(3-aminophenoxy)aniline
Synonyms: 4-(3-aminophenoxy)aniline
Molecular Weight (g/mol): 200.24
Molecular Formula: C12H12N2O
InChi Key: ZBMISJGHVWNWTE-UHFFFAOYSA-N
SMILES: NC1=CC=C(OC2=CC=CC(N)=C2)C=C1
Boiling Point 
  • (2)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)