accessibility menu, dialog, popup

UserName

Viewing profile as user:
CHOOCH

CAS RN 26627-36-1

IUPAC Name: 1,3-bis(prop-2-yn-1-yloxy)benzene
Synonyms: 1,3-bis(prop-2-yn-1-yloxy)benzene
Molecular Weight (g/mol): 186.21
Molecular Formula: C12H10O2
InChi Key: HSRDPYWXTZGWFQ-UHFFFAOYSA-N
SMILES: C#CCOC1=CC(OCC#C)=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)