accessibility menu, dialog, popup

UserName

Viewing profile as user:
NNNNNN

CAS RN 266349-83-1

IUPAC Name: 2-[4'-(4,6-diphenyl-1,3,5-triazin-2-yl)-[1,1'-biphenyl]-4-yl]-4,6-diphenyl-1,3,5-triazine
Synonyms: 2-[4'-(4,6-diphenyl-1,3,5-triazin-2-yl)-[1,1'-biphenyl]-4-yl]-4,6-diphenyl-1,3,5-triazine
Molecular Weight (g/mol): 616.73
Molecular Formula: C42H28N6
InChi Key: UICMBMCOVLMLIE-UHFFFAOYSA-N
SMILES: C1=CC=C(C=C1)C1=NC(=NC(=N1)C1=CC=CC=C1)C1=CC=C(C=C1)C1=CC=C(C=C1)C1=NC(=NC(=N1)C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results