accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COOOH

CAS RN 2664-60-0

IUPAC Name: dodecyl 4-hydroxybenzoate
Synonyms: dodecyl 4-hydroxybenzoate
Molecular Weight (g/mol): 306.45
Molecular Formula: C19H30O3
InChi Key: BAYSQTBAJQRACX-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCOC(=O)C1=CC=C(O)C=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)