accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2664-63-3

IUPAC Name: 4-[(4-hydroxyphenyl)sulfanyl]phenol
Synonyms: 4,4'-thiodiphenol 4,4'-sulfanediyldiphenol DFS p,p'-dihydroxydiphenyl sulfide bis(p-hydroxhphenyl) sulfide thiobisphenol 4,4'-dioxydiphenyl sulfide 4,4'-dioxydiphenylsulfide 4,4'-dihydroxydiphenyl sulfide 4,4'-dihydroxydiphenylsulfide
Molecular Weight (g/mol): 218.27
Molecular Formula: C12H10O2S
InChi Key: VWGKEVWFBOUAND-UHFFFAOYSA-N
SMILES: OC1=CC=C(SC2=CC=C(O)C=C2)C=C1
Percent Purity 
  • (2)
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)