accessibility menu, dialog, popup

UserName

Viewing profile as user:
NH2(R)(R)HOO

CAS RN 26685-83-6

IUPAC Name: (1R,2R)-2-aminocyclohexane-1-carboxylic acid
Synonyms: (1R,2R)-2-aminocyclohexane-1-carboxylic acid
Molecular Weight (g/mol): 143.19
Molecular Formula: C7H13NO2
InChi Key: USQHEVWOPJDAAX-PHDIDXHHSA-N
SMILES: N[C@@H]1CCCC[C@H]1C(O)=O
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)