accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3CH3H3COONHHO

CAS RN 26690-80-2

IUPAC Name: tert-butyl N-(2-hydroxyethyl)carbamate
Synonyms: tert-butyl N-(2-hydroxyethyl)carbamate
Molecular Weight (g/mol): 161.2
Molecular Formula: C7H15NO3
InChi Key: GPTXCAZYUMDUMN-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)NCCO
Molecular Weight (g/mol) 
  • (2)
  • (2)
  • (4)
Quantity 
  • (1)
  • (3)
  • (1)
  • (3)
Percent Purity 
  • (2)
  • (2)
  • (2)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (2)
  • (4)
  • (2)
  • (2)
  • (4)
  • (1)
  • (3)
  • (1)
  • (3)
  • (2)
  • (2)
  • (2)
  • (2)