accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 26741-53-7

IUPAC Name: 3,9-bis(2,4-di-tert-butylphenoxy)-2,4,8,10-tetraoxa-3,9-diphosphaspiro[5.5]undecane
Synonyms: weston 626
Molecular Weight (g/mol): 604.71
Molecular Formula: C33H50O6P2
InChi Key: AIBRSVLEQRWAEG-FJGSWBKINA-N
SMILES: CC(C)(C)C1=CC=C(O[P@]2OC[C@]3(CO[P@](OC4=CC=C(C=C4C(C)(C)C)C(C)(C)C)OC3)CO2)C(=C1)C(C)(C)C
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results