accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2675-89-0

CH3NCH3OCl

CAS RN 2675-89-0

IUPAC Name: 2-chloro-N,N-dimethylacetamide
Synonyms: 2-chloro-N,N-dimethylacetamide
Molecular Weight (g/mol): 121.56
Molecular Formula: C4H8ClNO
InChi Key: XBPPLECAZBTMMK-UHFFFAOYSA-N
SMILES: CN(C)C(=O)CCl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results