accessibility menu, dialog, popup

UserName

Viewing profile as user:
OOHOOHOHHO

CAS RN 26776-70-5

IUPAC Name: 2,5-bis(hydroxymethyl)-1,4-dioxane-2,5-diol
Synonyms: 2,5-bis(hydroxymethyl)-1,4-dioxane-2,5-diol
Molecular Weight (g/mol): 180.16
Molecular Formula: C6H12O6
InChi Key: KEQUNHIAUQQPAC-UHFFFAOYNA-N
SMILES: OCC1(O)COC(O)(CO)CO1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results