CAS RN 26782-74-1
IUPAC Name:
2-chloro-3-methylbutanoic acid
Synonyms:
2-chloro-3-methylbutanoic acid
Molecular Weight (g/mol):
136.58
Molecular Formula:
C5H9ClO2
InChi Key:
DDTJFSPKEIAZAM-UHFFFAOYNA-N
SMILES:
CC(C)C(Cl)C(O)=O
1
–
1
of
1
results
1
(S)-2-Chloro-3-methylbutyric Acid 98.0+%, TCI America™
CAS: 26782-74-1 Molecular Formula: C5H9ClO2 Molecular Weight (g/mol): 136.575 MDL Number: MFCD00067109 InChI Key: DDTJFSPKEIAZAM-BYPYZUCNSA-N Synonym: (S)-2-Chloroisovaleric Acid PubChem CID: 5324925 IUPAC Name: (2S)-2-chloro-3-methylbutanoic acid SMILES: CC(C)C(C(=O)O)Cl