accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 26787-75-7

IUPAC Name: 2-{[(benzyloxy)carbonyl]amino}-2-(4-hydroxyphenyl)acetic acid
Synonyms: {[(benzyloxy)carbonyl]amino}(4-hydroxyphenyl)acetic acid
Molecular Weight (g/mol): 301.3
Molecular Formula: C16H15NO5
InChi Key: CGDQNVNWDDEJPY-UHFFFAOYNA-N
SMILES: OC(=O)C(NC(=O)OCC1=CC=CC=C1)C1=CC=C(O)C=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)