accessibility menu, dialog, popup

UserName

Viewing profile as user:
ClH3CNCH3NNCH3CH3SNOO

CAS RN 2679-01-8

IUPAC Name: 3,7-bis(dimethylamino)-4-nitro-5λ⁴-phenothiazin-5-ylium chloride
Synonyms: methylene green
Molecular Weight (g/mol): 364.85
Molecular Formula: C16H17ClN4O2S
InChi Key: YYGBVRCTHASBKD-UHFFFAOYSA-M
SMILES: [Cl-].CN(C)C1=CC=C2N=C3C=CC(N(C)C)=C(C3=[S+]C2=C1)[N+]([O-])=O
Color 
  • (2)
  • (3)
Grade 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)