accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NCH3OH2C

CAS RN 2680-03-7

IUPAC Name: N,N-dimethylprop-2-enamide
Synonyms: N,N-dimethylacrylamide
Molecular Weight (g/mol): 99.13
Molecular Formula: C5H9NO
InChi Key: YLGYACDQVQQZSW-UHFFFAOYSA-N
SMILES: CN(C)C(=O)C=C
Boiling Point 
  • (2)
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (5)
  • (2)
  • (2)
  • (3)
  • (2)
  • (2)
  • (3)
  • (7)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)