accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 26815-13-4

IUPAC Name: 3-[3-(dimethylamino)propoxy]benzaldehyde
Synonyms: 3-[3-(dimethylamino)propoxy]benzaldehyde
Molecular Weight (g/mol): 207.27
Molecular Formula: C12H17NO2
InChi Key: CMVPGTWBRBJOQR-UHFFFAOYSA-N
SMILES: CN(C)CCCOC1=CC=CC(C=O)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results