accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 26829-77-6

IUPAC Name: 3-chloro-2,6-dimethylaniline
Synonyms: 3-chloro-2,6-dimethylaniline
Molecular Weight (g/mol): 155.63
Molecular Formula: C8H10ClN
InChi Key: FGMXFOTYCHZCLA-UHFFFAOYSA-N
SMILES: CC1=CC=C(Cl)C(C)=C1N
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)