accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 2683-70-7

IUPAC Name: 4-(phenoxymethyl)benzaldehyde
Synonyms: 4-(phenoxymethyl)benzaldehyde
Molecular Weight (g/mol): 212.25
Molecular Formula: C14H12O2
InChi Key: JLGXYDMVIJFOKF-UHFFFAOYSA-N
SMILES: O=CC1=CC=C(COC2=CC=CC=C2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results