accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 26864-56-2

IUPAC Name: 1-[4,4-bis(4-fluorophenyl)butyl]-4-[4-chloro-3-(trifluoromethyl)phenyl]piperidin-4-ol
Synonyms: penfluridol semap janssen brand of penfluridol C28H27ClF5NO penfluridolum
Molecular Weight (g/mol): 523.97
Molecular Formula: C28H27ClF5NO
InChi Key: MDLAAYDRRZXJIF-UHFFFAOYSA-N
SMILES: OC1(CCN(CCCC(C2=CC=C(F)C=C2)C2=CC=C(F)C=C2)CC1)C1=CC=C(Cl)C(=C1)C(F)(F)F
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results