accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3CNO

CAS RN 2687-91-4

IUPAC Name: 1-ethylpyrrolidin-2-one
Synonyms: ethyl pyrrolidone
Molecular Weight (g/mol): 113.16
Molecular Formula: C6H11NO
InChi Key: ZFPGARUNNKGOBB-UHFFFAOYSA-N
SMILES: CCN1CCCC1=O
Boiling Point 
  • (3)
  • (2)
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (8)
Quantity 
  • (3)
  • (2)
  • (1)
  • (1)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (2)
  • (2)
  • (6)
  • (3)
  • (2)
  • (3)
  • (2)
  • (2)
  • (8)
  • (3)
  • (2)
  • (1)
  • (1)
  • (3)
  • (5)