accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 268730-86-5

IUPAC Name: (2S)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-5-({4-[(E)-[1-(2-hydroxy-4,4-dimethyl-6-oxocyclohex-1-en-1-yl)-3-methylbutylidene]amino]phenyl}methoxy)-5-oxopentanoic acid
Synonyms: (2S)-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}-5-({4-[(E)-[1-(2-hydroxy-4,4-dimethyl-6-oxocyclohex-1-en-1-yl)-3-methylbutylidene]amino]phenyl}methoxy)-5-oxopentanoic acid
Molecular Weight (g/mol): 680.8
Molecular Formula: C40H44N2O8
InChi Key: BHDQEOLPKMRQEQ-GQVZARDDSA-N
SMILES: CC(C)C\C(=N/C1=CC=C(COC(=O)CC[C@H](NC(=O)OCC2C3=C(C=CC=C3)C3=C2C=CC=C3)C(O)=O)C=C1)C1=C(O)CC(C)(C)CC1=O
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)