accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 26889-39-4

IUPAC Name: 1-[3-amino-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-1,2,3,4-tetrahydropyrimidine-2,4-dione
Synonyms: 1-[3-amino-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-3H-pyrimidine-2,4-dione
Molecular Weight (g/mol): 243.22
Molecular Formula: C9H13N3O5
InChi Key: LLIPTMWIZVIUSX-UHFFFAOYNA-N
SMILES: NC1C(O)C(CO)OC1N1C=CC(=O)NC1=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results