CAS RN 2689-69-2
CAS RN 2689-69-2
IUPAC Name:
methyl 3-oxothiolane-2-carboxylate
Synonyms:
methyl 3-oxothiolane-2-carboxylate
Molecular Weight (g/mol):
160.19
Molecular Formula:
C6H8O3S
InChi Key:
TUSSXVYKOOVOID-UHFFFAOYNA-N
SMILES:
COC(=O)C1SCCC1=O
1
–
1
of
1
results
1
Sigma Aldrich Methyl 3-oxotetrahydrothiophene-2-carboxylate
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.