accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 26893-68-5

IUPAC Name: 6-nitropyridine-2-carboxylic acid
Synonyms: 6-nitropyridine-2-carboxylic acid
Molecular Weight (g/mol): 168.11
Molecular Formula: C6H4N2O4
InChi Key: MIHFOFLNJIPDRY-UHFFFAOYSA-N
SMILES: OC(=O)C1=NC(=CC=C1)[N+]([O-])=O
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results