accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 26929-65-7

IUPAC Name: 1-[3-azido-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-1,2,3,4-tetrahydropyrimidine-2,4-dione
Synonyms: 1-[3-azido-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-3H-pyrimidine-2,4-dione
Molecular Weight (g/mol): 269.22
Molecular Formula: C9H11N5O5
InChi Key: MRUKYOQQKHNMFI-UHFFFAOYNA-N
SMILES: OCC1OC(C(N=[N+]=[N-])C1O)N1C=CC(=O)NC1=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results