accessibility menu, dialog, popup

UserName

Viewing profile as user:
H2N

CAS RN 2693-46-1

IUPAC Name: fluoranthen-3-amine
Synonyms: 3-fluoranthenamine
Molecular Weight (g/mol): 217.27
Molecular Formula: C16H11N
InChi Key: VHGJAFIHUSTRGB-UHFFFAOYSA-N
SMILES: NC1=C2C=CC=C3C4=CC=CC=C4C(C=C1)=C23
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results