CAS RN 269390-77-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MilliporeSigma™ Calbiochem™ VEGFR Tyrosine Kinase Inhibitor VI, AAL-993
The VEGFR Tyrosine Kinse Inhibitor VI, ALL-993, also referenced under CAS 269390-77-4, controls the biological activity of VEGFR Tyrosine Kinase. This small molecule/inhibitor is primarily used for Cancer applications.