accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 269409-97-4

IUPAC Name: 2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol
Synonyms: 2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol
Molecular Weight (g/mol): 220.08
Molecular Formula: C12H17BO3
InChi Key: VLROJECCXBBKPZ-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CC=CC=C1O
Boiling Point 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results