accessibility menu, dialog, popup

UserName

Viewing profile as user:
BOOH3CH3CCH3H3COH

CAS RN 269410-21-1

IUPAC Name: 6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)naphthalen-2-ol
Synonyms: 6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)naphthalen-2-ol
Molecular Weight (g/mol): 270.14
Molecular Formula: C16H19BO3
InChi Key: FEOQRVDXBOSJLZ-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CC=C2C=C(O)C=CC2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results