accessibility menu, dialog, popup

UserName

Viewing profile as user:
NN

CAS RN 26947-41-1

IUPAC Name: isoquinoline-3-carbonitrile
Synonyms: isoquinoline-3-carbonitrile
Molecular Weight (g/mol): 154.17
Molecular Formula: C10H6N2
InChi Key: GXXBVBVNGMCFIU-UHFFFAOYSA-N
SMILES: N#CC1=CC2=CC=CC=C2C=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results