accessibility menu, dialog, popup

UserName

Viewing profile as user:
NaOSOOCH2

CAS RN 2695-37-6

IUPAC Name: sodium 4-ethenylbenzene-1-sulfonate
Synonyms: sodium 4-styrenesulfonate
Molecular Weight (g/mol): 206.19
Molecular Formula: C8H7NaO3S
InChi Key: XFTALRAZSCGSKN-UHFFFAOYSA-M
SMILES: [Na+].[O-]S(=O)(=O)C1=CC=C(C=C)C=C1
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)
  • (2)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)