accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 269730-03-2

IUPAC Name: 3-[(4-hydroxyphenyl)methylidene]-5,6-dimethoxy-2,3-dihydro-1H-indol-2-one
Synonyms: 3-[(4-hydroxyphenyl)methylidene]-5,6-dimethoxy-1H-indol-2-one
Molecular Weight (g/mol): 297.31
Molecular Formula: C17H15NO4
InChi Key: JGSMCYNBVCGIHC-UHFFFAOYSA-N
SMILES: COC1=C(OC)C=C2C(NC(=O)C2=CC2=CC=C(O)C=C2)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results