accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 27048-04-0

IUPAC Name: 6-chloro-3-nitropyridin-2-amine
Synonyms: 6-chloro-3-nitropyridin-2-amine
Molecular Weight (g/mol): 173.56
Molecular Formula: C5H4ClN3O2
InChi Key: WERABQRUGJIMKQ-UHFFFAOYSA-N
SMILES: NC1=NC(Cl)=CC=C1[N+]([O-])=O
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results